Flexible inspection light
Lieferant: Restek
22627
REST22627EA 73 EUR
REST22627
Flexible inspection light
Beleuchtung LED-Beleuchtung
This flexible inspeciton light inspects inside surfaces of sample cylinders or other chambers.
  • Has a reach of 355 mm
  • LED life of 100000 hours