Injection port weldments
Lieferant: Restek
22664 22665 22666 22667 22668 22669
REST22664EA 650 EUR
REST22664 REST22665 REST22666 REST22667 REST22668 REST22669
Injection port weldments
Chromatographie-Zubehör Chromatographie-Ersatzteile
Injection port weldments for Agilent GCs for use with purge and trap systems. This can easily attach your purge and trap with pre-installed low dead volume fittings.