Rough pump oil
Lieferant: Restek
22687
REST22687EA 48 EUR
REST22687
Rough pump oil
Schmiermittel
This rough pump oil ideal for MSD and oil vacuum pumps is formulated from crude oil stocks known for their durability and line-lubricating qualities.
It can be used in Agilent 5973/5972/5971 and GCD mass spec systems or in other MSD systems that require rough pump oil.