Zero dilution inner liner
Lieferant: Restek
22991
REST22991EA 207 EUR
REST22991
Zero dilution inner liner
Chromatographie-Zubehör Chromatographie-Liner
This liner is ideal for PerkinElmer auto SYS and clarus 580/680 GCs equipped with split/splitless inlets.
Made from borosilicate glass.