Tubing cutter, Ridgid®
Lieferant: Restek
23011
REST23011EA 66 EUR
REST23011
Tubing cutter, Ridgid®
Schneidewerkzeuge
These compact cutters are suitable for cutting metal tubings.
  • Replaceable cutting wheel
  • Not for use with stainless steel tubing