Rethreading tool
Lieferant: Restek
23016 23017 23018
REST23016EA 109 EUR
REST23016 REST23017 REST23018
Rethreading tool
Werkzeugsätze
These rethreading tools are suitable for repairing worn or damaged threads that come as a result of consistent use and temperature changes.
  • Multiple uses (injection ports, fittings, etc.)
  • Built-in guide prevents cross-threading
Repeated installation, removal and temperature changes cause wear and damage to threads on GC parts. This can cause a poor seal, and oxygen can enter the system, compromising analytical results and possibly destroying expensive analytical columns.