Tube cutting machine, SSI TC-20
Lieferant: Restek
23029
REST23029EA 2740 EUR
REST23029
Tube cutting machine, SSI TC-20
Schneidewerkzeuge
This tube cutting maching cuts ¹/₁₆", ¹/₈" or ¹/₄" tubing with inside diameter as small as 0,008".
  • Electrically operated bench-top model
  • Handy dressing tool on the swing arm removes burrs and reams tubing
  • Voltage selectable 110 to 120/220 to 240 volts, 50 to 60 Hz