Inlet liners, topaz
Lieferant: Restek
23325 23278 23279 23280 23281 23282 23283 23284 23285 23286 23287 23288 23289 23291 23333 23293 23294 23295 23296 23297 23300 23301 23302 23303 23306 23307 23308 23309 23310 23311 23313 23314 23315 23316 23317 23318 23319 23320 23321 23322 23323 23324 23327 23328 23329 23330 23331 23332 23336 23428 23429 23430 23431 23432 23434 23435 23436 23437 23438 23439 23440 23441 23442 23443 23444 23445 89023446 23447 23448 23449 23450 23451 23452 23453 23454 23455 23456 23457 23459 23460 23461 23462 23463 23465 23466 23467 23468 23471 23472
REST23325EA 263 EUR
REST23325 REST23278 REST23279 REST23280 REST23281 REST23282 REST23283 REST23284 REST23285 REST23286 REST23287 REST23288 REST23289 REST23291 REST23333 REST23293 REST23294 REST23295 REST23296 REST23297 REST23300 REST23301 REST23302 REST23303 REST23306 REST23307 REST23308 REST23309 REST23310 REST23311 REST23313 REST23314 REST23315 REST23316 REST23317 REST23318 REST23319 REST23320 REST23321 REST23322 REST23323 REST23324 REST23327 REST23328 REST23329 REST23330 REST23331 REST23332 REST23336 REST23428 REST23429 REST23430 REST23431 REST23432 REST23434 REST23435 REST23436 REST23437 REST23438 REST23439 REST23440 REST23441 REST23442 REST23443 REST23444 REST23445 REST89023446 REST23447 REST23448 REST23449 REST23450 REST23451 REST23452 REST23453 REST23454 REST23455 REST23456 REST23457 REST23459 REST23460 REST23461 REST23462 REST23463 REST23465 REST23466 REST23467 REST23468 REST23471 REST23472
Inlet liners, topaz
Chromatographie-Zubehör Chromatographie-Liner
Topaz GC inlet liners feature revolutionary technology and inertness.
  • Low breakdown for accurate and precise low-level GC analyses
  • Strict manufacturing controls and QC testing for superior reliability across compound classes
  • High cleanliness for maximised GC uptime and lab throughput