Humidification chamber
Lieferant: Restek
24282
REST24282EA 536 EUR
REST24282
Humidification chamber
Probenehmer Luftkeimsammler
It is important to use humidified air or nitrogen to help remove volatile organic contaminants when cleaning SilcoCan® or TO-Can® canisters.
  • Chamber withstands pressure up to 90 psi
  • Easy-to-open lid for filling with water
  • Inlet and outlet compression fittings for easy connection to pressure lines on your cleaning system
It is important to use humidified air or nitrogen to help remove volatile organic contaminants when cleaning SilcoCan® or TO-Can® canisters.