ValvTool wrench
Lieferant: Restek
25321
REST25321EA 58 EUR
REST25321
ValvTool wrench
Schraubenschlüssel
The ValvTool is a time saving device that provides easy access to many hard-to-reach Rheodyne or Valco valves.