Solid phase extraction cartridges, CarboPrep®
Lieferant: Restek
26206
REST26206EA 0 EUR
REST26206
Solid phase extraction cartridges, CarboPrep®
Chromatographiesäulen
The reversing cartridge design allows convenient inverted elution of strongly retained analytes using minimum solvent volumes. Ideal for extraction of pesticides in water.
  • High adsorbent capacity (surface area ~200 m²/g) for large volume sampling
Chromatographic grade graphitised carbon provides consistent and quantitative recoveries of a wide variety of semivolatiles, pesticides and herbicides.