High purity airgas VOC regulators
Lieferant: Restek
26371 26372
REST26371EA 2460 EUR
REST26371 REST26372
High purity airgas VOC regulators
Druckminderer
These single stage stainless steel VOC regulators features stainless steel diaphragm, two pressure gauges, CGA-180 fitting and Kel-F® seat.
  • Accurate pressure control even at low flow rates
  • Individually tested for leaks and impurities