VOA Surrogate spike mix (3 components)
Lieferant: Restek
30004
REST30004EA
51
EUR
REST30004
VOA Surrogate spike mix (3 components)
Standards
Organische Standards
Mix contains: 1-Bromo-4-fluorobenzene (BFB) (460-00-4), 1,2-Dichloroethane-d4 (17060-07-0) and Toluene-d8 (2037-26-5).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.