VOA calibration mix #1 (ketones) (4 components)
Lieferant: Restek
30006
REST30006EA
58
EUR
REST30006
VOA calibration mix #1 (ketones) (4 components)
Standards
Organische Standards
Mix contains Acetone (67-64-1), 2-Butanone (MEK) (78-93-3), 2-Hexanone (591-78-6), 4-Methyl-2-pentanone (MIBK) (108-10-1).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.