VOA calibration mix #3 (10 components)
Lieferant: Restek
30008
REST30008EA
52
EUR
REST30008
VOA calibration mix #3 (10 components)
Standards
Organische Standards
Mix contains: Carbon tetrachloride (56-23-5), Chlorobenzene (108-90-7), Chloroform (67-66-3), 1,1-Dichloroethane (75-34-3), 1,1-Dichloroethene (75-35-4), 1,2-Dichloropropane (78-87-5), Methylene chloride (dichloromethane) (75-09-2), 1,1,2-Trichloroethane (79-00-5), Trichloroethene (79-01-6), m-Xylene (108-38-3).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.