8021 Surrogate mix (2 components)
Lieferant: Restek
30086
REST30086EA
58
EUR
REST30086
8021 Surrogate mix (2 components)
Standards
Organische Standards
Mix contains: 2-Bromochlorobenzene (694-80-4), 1,4-Dichlorobutane (110-56-5).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.