8260B MegaMix calibration mix, revised (75 components)
Lieferant: Restek
The mix consists of acetonitrile, acrylonitrile, allyl chloride , benzene, bromobenzene, bromochloromethane, bromodichloromethane, bromoform, n-butylbenzene, sec-butylbenzene, tert-butylbenzene, carbon disulfide, carbon tetrachloride, chlorobenzene, 2-chloroethanol, chloroform, chloroprene , 2-chlorotoluene, 4-chlorotoluene, dibromochloromethane, 1,2-dibromo-3-chloropropane , 1,2-dibromoethane , dibromomethane, 1,2-dichlorobenzene, 1,3-dichlorobenzene, 1,4-dichlorobenzene, cis-1,4-dichloro-2-butene, trans-1,4-dichloro-2-butene, 1,1-dichloroethane, 1,2-dichloroethane, 1,1-dichloroethene, cis-1,2-dichloroethene, trans-1,2-dichloroethene, 1,2-dichloropropane, 1,3-dichloropropane, 2,2-dichloropropane, 1,1-dichloropropene, cis-1,3-dichloropropene, trans-1,3-dichloropropene, diethyl ether , 1,4-dioxane, ethylbenzene, ethyl methacrylate, hexachloro-1,3-butadiene, iodomethane , isobutyl alcohol , isopropylbenzene , 4-isopropyl toluene , methacrylonitrile, methyl acrylate, methyl methacrylate, methylene chloride , naphthalene, nitrobenzene, 2-nitropropane, propionitrile, n-propylbenzene, styrene, 1,1,1,2-tetrachloroethane, 1,1,2,2-tetrachloroethane, tetrachloroethene, tetrahydrofuran, toluene, 1,2,3-trichlorobenzene, 1,1,1-trichloroethane, 1,1,2-trichloroethane, trichloroethene, 1,2,3-trichloropropane, 1,1,2-trichlorotrifluororethane , 1,2,4-trimethylbenzene, 1,3,5-trimethylbenzene, m-xylene, o-xylene, p-xylene.
- Contains 75 compounds in one convenient ampul for the analysis of VOCs in solid waste
- Quantitatively analyzed to confirm concentration and identity of each compound
- Excludes pentachloroethane for labs experiencing elevated tetrachloroethene response caused by pentachloroethane breakdown
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.