524 Internal standard/surrogate mix (3 components)
Lieferant: Restek
30201
REST30201EA
58
EUR
REST30201
524 Internal standard/surrogate mix (3 components)
Standards
Organische Standards
Mix contains 1-Bromo-4-fluorobenzene (BFB) (460-00-4), 1,2-Dichlorobenzene-d4 (2199-69-1), Fluorobenzene (462-06-6).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.