Calibration mix #7 Kit, 524
Lieferant: Restek
30202
REST30202EA 70.5 EUR
REST30202
Calibration mix #7 Kit, 524
Standards Organische Standards
Contains 1 ml each of these mixtures: 524 calibration mix #7A, 524 calibration mix #7B. calibration mix #7A contains Acetone (67-64-1), 2-Butanone (MEK) (78-93-3), 2-Hexanone (591-78-6), 4-Methyl-2-pentanone (MIBK) (108-10-1), Tetrahydrofuran (109-99-9); calibration mix #7B contains Acrylonitrile (107-13-1)
Allyl chloride (3-chloropropene) (107-05-1), Ethyl methacrylate (97-63-2), Methyl acrylate (96-33-3), Methyl methacrylate (80-62-6), Nitrobenzene (98-95-3), Pentachloroethane (76-01-7).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.