VOA purgeable halocarbon mix #1 (23 components)
Lieferant: Restek
30212
REST30212EA 100 EUR
REST30212
VOA purgeable halocarbon mix #1 (23 components)
Standards Organische Standards
Mix contains: Bromodichloromethane (75-27-4); Bromoform (75-25-2); Carbon tetrachloride (56-23-5); Chlorobenzene (108-90-7); 2-Chloroethyl vinyl ether (110-75-8); Chloroform (67-66-3); Dibromochloromethane (124-48-1); 1,2-Dichlorobenzene (95-50-1); 1,3-Dichlorobenzene (541-73-1); 1,4-Dichlorobenzene (106-46-7); 1,1-Dichloroethane (75-34-3); 1,2-Dichloroethane (107-06-2); 1,1-Dichloroethene (75-35-4); trans-1,2-Dichloroethene (156-60-5); 1,2-Dichloropropane (78-87-5); cis-1,3-Dichloropropene (10061-01-5); trans-1,3-Dichloropropene (10061-02-6); Methylene chloride (dichloromethane) (75-09-2); 1,1,2,2-Tetrachloroethane (79-34-5); Tetrachloroethene (127-18-4); 1,1,1-Trichloroethane (71-55-6); 1,1,2-Trichloroethane (79-00-5); Trichloroethene (79-01-6).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.