8240 Alcohols mix (5 components)
Lieferant: Restek
30214
REST30214EA
57
EUR
REST30214
8240 Alcohols mix (5 components)
Standards
Organische Standards
Mix contains: Allyl alcohol (2-propen-1-ol) (107-18-6); 2-Chloroethanol (107-07-3); Ethanol (64-17-5); Isobutyl alcohol (78-83-1); Propargyl alcohol (107-19-7).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.