PVOC mix (California) (7 components)
Lieferant: Restek
30231
REST30231EA
62
EUR
REST30231
PVOC mix (California) (7 components)
Standards
Organische Standards
Mix contains: Benzene (71-43-2); Ethylbenzene (100-41-4); Methyl tert-butyl ether (MTBE) (1634-04-4); Toluene (108-88-3); m-Xylene (108-38-3); o-Xylene (95-47-6); p-Xylene (106-42-3).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.