CA WIP VOA standard (11 components)
Lieferant: Restek
30236
REST30236EA
71.5
EUR
REST30236
CA WIP VOA standard (11 components)
Standards
Mineralölindustrie-Standards
For monitoring samples suspected of gasoline contamination requires laboratories to calibrate and report these compounds.
It consists of benzene, chlorobenzene, 1,2-dichlorobenzene, 1,3-dichlorobenzene, 1,4-dichlorobenzene, ethylbenzene, methyl tert-butyl ether, toluene, m-xylene, o-xylene, p-xylene.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.