Certified BTEX in unleaded gas composite standard (9 components)
Lieferant: Restek
30237
REST30237EA
62
EUR
REST30237
Certified BTEX in unleaded gas composite standard (9 components)
Standards
Organische Standards
The BTEX in unleaded gas composite standard consists of 9 components, they are benzene, ethylbenzene, isopropyl benzene, methyl tert-butyl ether (MTBE), naphthalene, toluene, m-xylene, o-xylene and p-xylene.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.