504.1 Calibration mix (3 components)
Lieferant: Restek
30239
REST30239EA 40 EUR
REST30239
504.1 Calibration mix (3 components)
Standards Organische Standards
Mix contains: Chlorobenzene-d5 (3114-55-4); 1,4-Dichlorobenzene-d4 (3855-82-1); Fluorobenzene (462-06-6).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.