8260A surrogate mix (4 components)
Lieferant: Restek
30240
REST30240EA 65 EUR
REST30240
8260A surrogate mix (4 components)
Standards Umweltanalyse-Standards
The surrogate mix consists of four components 1-bromo-4-fluorobenzene (BFB), dibromofluoromethane, 1,2-dichloroethane-d4 and toluene-d8.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.