624 Surrogate standard mix (3 components)
Lieferant: Restek
30243
REST30243EA 58 EUR
REST30243
624 Surrogate standard mix (3 components)
Standards Organische Standards
Mix contains: 1-Bromo-4-fluorobenzene (BFB) (460-00-4); Fluorobenzene (462-06-6); Pentafluorobenzene (363-72-4).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.