524 calibration mix #7A (5 components)
Lieferant: Restek
The calibration mix consists of five components they are acetone, 2-butanone, 2-hexanone, 4-methyl-2-pentanone and tetrahydrofuran.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.