524 calibration mix #7B (7 components)
Lieferant: Restek
30304
REST30304EA
58
EUR
REST30304
524 calibration mix #7B (7 components)
Standards
Organische Standards
The calibration mix consists of seven components they are acrylonitrile, allyl chloride (3-chloropropene), ethyl methacrylate, methyl acrylate, methyl methacrylate, nitrobenzene and pentachloroethane.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.