Calibration check mix, 8240/8260 (6 components)
Lieferant: Restek
30427
REST30427EA 40 EUR
REST30427
Calibration check mix, 8240/8260 (6 components)
Standards Organische Standards
Mix contains chloroform, 1,1-dichloroethene, 1,2-dichloropropane, ethylbenzene, toluene, vinyl chloride.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.