PA DEP UST standard (11 components)
Lieferant: Restek
30433
REST30433EA 69.5 EUR
REST30433
PA DEP UST standard (11 components)
Standards Mineralölindustrie-Standards
Standard contains benzene, 1,2-dibromoethane (EDB), 1,2-dichloroethane, ethylbenzene (100-41-4), isopropyl benzene (cumene), methyl tert-butyl ether (MTBE), naphthalene, toluene, m-xylene, o-xylene, p-xylene.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.