502.2 Calibration mix #1 (gases) (6 components)
Lieferant: Restek
30439
REST30439EA 82.5 EUR
REST30439
502.2 Calibration mix #1 (gases) (6 components)
Standards Umweltanalyse-Standards
Calibration mix contains bromomethane (methyl bromide), chloroethane (ethyl chloride), chloromethane (methyl chloride), dichlorodifluoromethane (CFC-12), trichlorofluoromethane (CFC-11), vinyl chloride.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.