CLP 04.1 VOA internal standard/SMC spike mix (6 components)
Lieferant: Restek
30457
REST30457EA 63 EUR
REST30457
CLP 04.1 VOA internal standard/SMC spike mix (6 components)
Standards Organische Standards
The VOA internal standard/SMC spike mix consists of 6 components, they are bromochloromethane,1-bromo-4-fluorobenzene (BFB), chlorobenzene-d5, 1,2-dichloroethane-d4, 1,4-difluorobenzene and toluene-d8.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.