Alaska UST method AK101AA
Lieferant: Restek
30461
REST30461EA 70 EUR
REST30461
Alaska UST method AK101AA
Standards Mineralölindustrie-Standards
The alaska UST method consists of 14 components they are benzene, ethylbenzene, 1-ethyl-2-methylbenzene, 1-ethyl-3-methylbenzene, 1-ethyl-4-methylbenzene, isopropylbenzene (cumene), n-propylbenzene, toluene, 1,2,3-trimethylbenzene, 1,2,4-trimethylbenzene, 1,3,5-trimethylbenzene, m-xylene, o-xylene and p-xylene.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.