California oxygenates mix (5 components)
Lieferant: Restek
30465
REST30465EA
65
EUR
REST30465
California oxygenates mix (5 components)
Standards
Umweltanalyse-Standards
The california oxygenates mix consists of five components they are tert-amyl methyl ether (TAME) 2000 μg/ml, tert-butanol (TBA) 10000 μg/ml, diisopropyl ether (DIPE) 2000 μg/ml, ethyl-tert-butyl ether (ETBE) 2000 μg/ml and methyl tert-butyl ether (MTBE) 2000 μg/ml.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.