Michigan GRO mix (14 components)
Lieferant: Restek
30468
REST30468EA
57
EUR
REST30468
Michigan GRO mix (14 components)
Standards
Mineralölindustrie-Standards
It consists of benzene, 1,2-dibromoethane, 1,2-dichloroethane, ethylbenzene, isopropylbenzene (cumene), 2-methylnaphthalene, methyl tert-butyl-ether (MTBE), naphthalene, toluene, 1,2,4-trimethylbenzene, 1,3,5-trimethylbenzene, m-xylene, o-xylene and p-xylene.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.