Glycols standard
Lieferant: Restek
30471
REST30471EA
66
EUR
REST30471
Glycols standard
Standards
Mineralölindustrie-Standards
The glycol standard consists of two components they are ethylene glycol and propylene glycol.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.