Certified aromatics in gasoline, 16 components
Lieferant: Restek
Confirms unleaded gasoline TPH, BTEX and aromatics in single analysis.
This standard consists of 16 components.
Components: Benzene, ethylbenzene, m-ethyltoluene, o-ethyltoluene, p-ethyltoluene, isopropylbenzene, methyl tert-butyl ether (MTBE), naphthalene, n-propylbenzene, toluene, 1,2,3-trimethylbenzene, 1,2,4-trimethylbenzene, 1,3,5-trimethylbenzene, m-xylene, o-xylene, p-xylene.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.