Gasoline component standard, 10 components
Lieferant: Restek
30486
REST30486EA 52 EUR
REST30486
Gasoline component standard, 10 components
Standards Mineralölindustrie-Standards
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Gasoline component standard contains 10 components.
Components: Benzene (500 µg/ml), ethylbenzene (500 µg/ml), heptane (500 µg/ml), 2-methylpentane (1500 µg/ml), toluene (1500 µg/ml), 1,2,4-trimethylbenzene (1500 µg/ml), 2,2,4-trimethylpentane (1500 µg/ml), m-xylene (1000 µg/ml), o-xylene (1000 µg/ml) and p-xylene (1000 µg/ml).