NJDEP EPH 10/08 Rev. 2 aliphatics calibration standard (20 components)
Lieferant: Restek
30540
REST30540EA 70 EUR
REST30540
NJDEP EPH 10/08 Rev. 2 aliphatics calibration standard (20 components)
Standards Organische Standards
Consists of 20 components, those are n-nonane, n-decane, n-dodecane, n-tetradecane, n-hexadecane, n-octadecane, n-eicosane, n-heneicosane, n-docosane, n-tetracosane, n-hexacosane, n-octacosane, n-triacontane, n-dotriacontane, n-tetratriacontane, n-hexatriacontane, n-octatriacontane, n-tetracontane, 2-methylnaphthalene and naphthalene.
20 components
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.