NJDEP EPH 10/08 rev. 2 aliphatics matrix spike mix (18 components)
Lieferant: Restek
Consists of 18 components, those are n-nonane, n-decane, n-dodecane, n-tetradecane, n-hexadecane, n-octadecane, n-eicosane, n-heneicosane, n-docosane, n-tetracosane, n-hexacosane, n-octacosane, n-triacontane, n-dotriacontane, n-tetratriacontane, n-hexatriacontane, n-octatriacontane and n-tetracontane.
18 components
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.