NJDEP EPH 10/08 rev. 2 aromatics matrix spike mix (18 components)
Lieferant: Restek
30543
REST30543EA
144
EUR
REST30543
NJDEP EPH 10/08 rev. 2 aromatics matrix spike mix (18 components)
Standards
Organische Standards
Consists of 18 components, those are acenaphthene, acenaphthylene, anthracene, benzanthracene, benzopyrene, benzofluoranthene, benzoperylene, benzofluoranthene, chrysene, dibenzanthracene, fluoranthene, fluorene, indenopyrene, phenanthrene, 1,2,3-trimethylbenzene and pyrene.
18 components
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.