NJDEP EPH 10/08 rev. 2 aromatics fractionation check mix (18 components)
Lieferant: Restek
30545
REST30545EA
225
EUR
REST30545
NJDEP EPH 10/08 rev. 2 aromatics fractionation check mix (18 components)
Standards
Organische Standards
Consists of 18 components, those are acenaphthene, acenaphthylene, anthracene, benzanthracene, benzopyrene, benzofluoranthene, benzoperylene, benzofluoranthene, chrysene, dibenzanthracene, fluoranthene, fluorene, indenopyrene, 2-methylnaphthalene, naphthalene, phenanthrene, 1,2,3-trimethylbenzene and pyrene
16 components
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.