Ethylene oxide standard
Lieferant: Restek
Used as reference standards for detecting ethylene oxide and 1,4-dioxane in PEG-based products.
- Formulated with concentrations needed by labs analysing cosmetics and personal care products for possible carcinogens
- Quantitatively tested to confirm composition; detailed support documentation provided
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.