Ketones mix, 524.2 rev. 4.1 (5 components)
Lieferant: Restek
The ketones mix consists of five components they are acetone, 2-butanone (MEK), 1,1-dichloro-2-propanone, 2-hexanone and 4-methyl-2-pentanone (MIBK).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.