MA VPH standard with surrogate rev. 1.1 (July 2004) (16 components)
Lieferant: Restek
30604
REST30604EA
64.5
EUR
REST30604
MA VPH standard with surrogate rev. 1.1 (July 2004) (16 components)
Standards
Mineralölindustrie-Standards
The mix consists of (C5) n-pentane, (C9) n-nonane, (C10) n-decane, benzene, n-butylcyclohexane, 2,5-dibromotoluene, ethylbenzene, 2-methylpentane, methyl tert-butyl ether, naphthalene, toluene, 1,2,4-trimethylbenzene, 2,2,4-trimethylpentane, m-xylene, o-xylene, p-xylene.
- Standard for current Massachusetts VPH method - surrogate included
- More analyses per ampul-standard with surrogate is 10000 μg/ml
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.