MA VPH matrix spike mix with surrogate rev. 1.1 (July 2004) (16 components)
Lieferant: Restek
30605
REST30605EA 64.5 EUR
REST30605
MA VPH matrix spike mix with surrogate rev. 1.1 (July 2004) (16 components)
Standards Mineralölindustrie-Standards
The mix consists of (C5) n-pentane, (C9) n-nonane, (C10) n-decane, benzene, n-butylcyclohexane, 2,5-dibromotoluene (SS), ethylbenzene, 2-methylpentane, methyl tert-butyl ether, naphthalene, toluene, 1,2,4-trimethylbenzene, 2,2,4-trimethylpentane, m-xylene, o-xylene, p-xylene.
  • For current Massachusetts VPH method - surrogate included
  • Matrix spike mix at 50 μg/ml, per updated method
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.