CLP VOA CAL2000 MegaMix® standard
Lieferant: Restek
30632
REST30632EA 105.5 EUR
REST30632
CLP VOA CAL2000 MegaMix® standard
Standards Organische Standards
This MegaMix standard contains 28 components, 2000 µg/ml each in P and T methanol, 1 ml/ampule.
Components: Benzene, bromodichloromethane, bromoform, carbon disulphide, carbon tetrachloride, chlorobenzene, chloroform, dibromochloromethane, 1,1-dichloroethane, 1,2-dichloroethane, 1,1-dichloroethene, cis-1,2-dichloroethene, trans-1,2-dichloroethene, 1,2-dichloropropane, cis-1,3-dichloropropene, trans-1,3-dichloropropene, ethylbenzene, methylene chloride, styrene, 1,1,2,2-tetrachloroethane, tetrachloroethene, toluene, 1,1,1-trichloroethane, 1,1,2-trichloroethane, trichloroethene, m-xylene, o-xylene, p-xylene.
In methanol solution, ketones can interact with other compounds, resulting in a methoxy addition to the ketone.1 The presence of halogenated compounds greatly speeds the reaction, limiting shelf-life to less than two months
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.