ASTM® D3606 calibration standard #1 without internal standard (2 components)
Lieferant: Restek
30647
REST30647EA 216 EUR
REST30647
ASTM® D3606 calibration standard #1 without internal standard (2 components)
Standards Mineralölindustrie-Standards
The mix consists of benzene (5% vol/vol), toluene (20% vol/vol).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.