ASTM D3606 calibration standard #3 without internal standard (2 components)
Lieferant: Restek
30649
REST30649EA
216
EUR
REST30649
ASTM D3606 calibration standard #3 without internal standard (2 components)
Standards
Mineralölindustrie-Standards
The mix consists of benzene (1,25% vol/vol), toluene (10% vol/vol).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.